Please login to your online account to display your discounted pricing

Rabbit anti-Slit2, Clone: SP122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17450405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17450405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Cross Reactivity Rat
Primary or Secondary Primary
Regulatory Status RUO
Format Purified
Quantity 0.5mL
Immunogen Synthetic peptide corresponding to C-terminus of human SLIT2 protein.
Clone SP122
Conjugate Unlabeled
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Purified
Isotype IgG
Gene ID (Entrez) 9353
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Host Species Rabbit
Antigen Slit2