Please login to your online account to display your discounted pricing

Rabbit anti-Slit2, Clone: SP122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17450401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17450401

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9353
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Purification Method Purified
Clone SP122
Antigen Slit2
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Primary or Secondary Primary
Reconstitution Protein A purified
Format Purified
Immunogen Synthetic peptide corresponding to C-terminus of human SLIT2 protein.
Isotype IgG