Please login to your online account to display your discounted pricing

Rabbit anti-S100B, Clone: SP127, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17451610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17451610

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 6285
Format Purified
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Rat
Host Species Rabbit
Quantity 1mL
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Regulatory Status RUO
Clone SP127
Reconstitution Protein A purified
Purification Method Purified
Conjugate Unlabeled
Antigen S100B
Primary or Secondary Primary
Immunogen Full length recombinant human S100 beta.
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.