Please login to your online account to display your discounted pricing

Rabbit anti-S100B, Clone: SP127, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17451605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17451605

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen S100B
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Host Species Rabbit
Immunogen Full length recombinant human S100 beta.
Isotype IgG
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6285
Clone SP127
Purification Method Purified
Regulatory Status RUO
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat