Please login to your online account to display your discounted pricing

Rabbit anti-S100B, Clone: SP127, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17451601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17451601

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen S100B
Format Purified
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Immunogen Full length recombinant human S100 beta.
Isotype IgG
Quantity 0.1mL
Gene ID (Entrez) 6285
Clone SP127
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Host Species Rabbit
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified