Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: S19-P, Novus Biologicals™

$332.66 - $332.66

Specifications

Antigen Renal Cell Carcinoma (gp200)
Clone S19-P
Immunogen Peptide derived from N-terminal sequence of human CD205.
Gene Accession No. O60449
Gene ID (Entrez) 4065
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13014601

 
novus biologicals canada
NBP13014601ML
0.1mL Each for $332.66
Description & Specifications

Specifications

Antigen Renal Cell Carcinoma (gp200)
Clone S19-P
Immunogen Peptide derived from N-terminal sequence of human CD205.
Gene Accession No. O60449
Gene ID (Entrez) 4065
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)