Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: S19-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014605

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Gene Accession No. O60449
Host Species Rabbit
Immunogen Peptide derived from N-terminal sequence of human CD205.
Isotype IgG
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 4065
Clone S19-P
Conjugate Unlabeled
Antigen Renal Cell Carcinoma (gp200)
Test Specificity This antibody is specific for the N terminus of CD205