Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: S19-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014610

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Antigen Renal Cell Carcinoma (gp200)
Format Affinity Purified
Isotype IgG
Purification Method Affinity Purified
Quantity 1mL
Regulatory Status RUO
Test Specificity This antibody is specific for the N terminus of CD205
Cross Reactivity Human
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4065
Clone S19-P
Conjugate Unlabeled
Gene Accession No. O60449
Immunogen Peptide derived from N-terminal sequence of human CD205.