Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: S19-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014601

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Immunogen Peptide derived from N-terminal sequence of human CD205.
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4065
Test Specificity This antibody is specific for the N terminus of CD205
Conjugate Unlabeled
Antigen Renal Cell Carcinoma (gp200)
Gene Accession No. O60449
Host Species Rabbit
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Clone S19-P