Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: R18-D, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13014710ML

 View all options for this product


For Research Use Only

Catalog No. NBP13014710


Description & Specifications


Antigen Renal Cell Carcinoma (gp200)
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone R18-D
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. O60449
Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human CD205.
Isotype IgG
Purification Method Affinity Purified
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4065
Test Specificity This antibody is specific for the C terminus of CD205