Please login to your online account to display your discounted pricing

Rabbit anti-Renal Cell Carcinoma (gp200), Clone: R18-D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014701

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Peptide derived from C-terminal sequence of human CD205.
Gene Accession No. O60449
Format Affinity Purified
Quantity 0.1mL
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Test Specificity This antibody is specific for the C terminus of CD205
Antigen Renal Cell Carcinoma (gp200)
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Host Species Rabbit
Clone R18-D
Regulatory Status RUO
Purification Method Affinity Purified
Gene ID (Entrez) 4065