Please login to your online account to display your discounted pricing

Rabbit anti-PTH, Clone: SP151, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251401

  • / Each
 

Description & Specifications

Specifications

Antigen PTH
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP151
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from the internal region of human PTH protein.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5741
Test Specificity Predicted to cross react with Bovine, Canine, Monkey, Porcine.
Reconstitution Protein A purified
Target Species Human