Please login to your online account to display your discounted pricing

Rabbit anti-PTH, Clone: SP151, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Immunogen Synthetic peptide derived from the internal region of human PTH protein.
Isotype IgG
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5741
Conjugate Unlabeled
Cross Reactivity Human
Antigen PTH
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Test Specificity Predicted to cross react with Bovine, Canine, Monkey, Porcine.
Reconstitution Protein A purified
Clone SP151
Purification Method Purified