Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: X21-C, Novus Biologicals™

$367.12 - $569.00

Specifications

Antigen Progesterone R/NR3C3
Clone X21-C
Immunogen Peptide derived from N-terminal region of human progesterone receptor.
Gene Accession No. P06401
Gene ID (Entrez) 5241
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17907720

 
novus biologicals canada
NBP179077200UL
200μL Each for $569.00

NBP17907710

 
novus biologicals canada
NBP179077100UL
100μL Each for $367.12
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone X21-C
Immunogen Peptide derived from N-terminal region of human progesterone receptor.
Gene Accession No. P06401
Gene ID (Entrez) 5241
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)