Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: X21-C, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179077200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907720

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Immunogen Peptide derived from N-terminal region of human progesterone receptor.
Isotype IgG
Antigen Progesterone R/NR3C3
Format Affinity Purified
Gene Accession No. P06401
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone X21-C
Conjugate Unlabeled
Cross Reactivity Human
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241