Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: X21-C, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179077100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907710

  • / Each
 

Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone X21-C
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P06401
Host Species Rabbit
Immunogen Peptide derived from N-terminal region of human progesterone receptor.
Isotype IgG
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241
Target Species Human