Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: SP2, Novus Biologicals™

$363.90 - $769.59

Specifications

Antigen Progesterone R/NR3C3
Clone SP2
Immunogen Recombinant protein encoding aa 412-526 of human progesterone receptor.
Gene ID (Entrez) 5241
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21246301

 
novus biologicals canada
NBP21246301ML
0.1mL Each for $363.90

NBP21246305

 
novus biologicals canada
NBP21246305ML
0.5mL Each for $769.59
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone SP2
Immunogen Recombinant protein encoding aa 412-526 of human progesterone receptor.
Gene ID (Entrez) 5241
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)