Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: SP2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21246305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21246305

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Cross Reactivity Human
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Format Supernatant
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Immunogen Recombinant protein encoding aa 412-526 of human progesterone receptor.
Quantity 0.5mL
Antigen Progesterone R/NR3C3
Isotype IgG
Test Specificity Predicted to cross react with Bovine, Porcine, Rabbit.
Purification Method Supernatant
Clone SP2
Gene ID (Entrez) 5241