Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: SP2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21246301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21246301

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Antigen Progesterone R/NR3C3
Format Supernatant
Gene ID (Entrez) 5241
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Clone SP2
Host Species Rabbit
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Recombinant protein encoding aa 412-526 of human progesterone receptor.
Regulatory Status RUO
Test Specificity Predicted to cross react with Bovine, Porcine, Rabbit.
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human