Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: P21-S, Novus Biologicals™

$367.12 - $569.00

Specifications

Antigen Progesterone R/NR3C3
Clone P21-S
Immunogen Peptide derived from C-terminus of inhibitory domain (IF) of human progesterone receptor.
Gene Accession No. P06401-1
Gene ID (Entrez) 5241
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17900410

 
novus biologicals canada
NBP179004100UL
100μL Each for $367.12

NBP17900420

 
novus biologicals canada
NBP179004200UL
200μL Each for $569.00
Description & Specifications

Specifications

Antigen Progesterone R/NR3C3
Clone P21-S
Immunogen Peptide derived from C-terminus of inhibitory domain (IF) of human progesterone receptor.
Gene Accession No. P06401-1
Gene ID (Entrez) 5241
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)