Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: P21-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179004200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17900420

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Cross Reactivity Human
Primary or Secondary Primary
Antigen Progesterone R/NR3C3
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS
Regulatory Status RUO
Clone P21-S
Conjugate Unlabeled
Isotype IgG
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Storage Requirements Store at 4°C. Do not freeze.
Immunogen Peptide derived from C-terminus of inhibitory domain (IF) of human progesterone receptor.
Quantity 200μL
Gene ID (Entrez) 5241
Gene Accession No. P06401-1
Monoclonal or Polyclonal Monoclonal