Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: P21-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179004100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17900410

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS
Host Species Rabbit
Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Antigen Progesterone R/NR3C3
Primary or Secondary Primary
Immunogen Peptide derived from C-terminus of inhibitory domain (IF) of human progesterone receptor.
Isotype IgG
Purification Method Affinity Purified
Clone P21-S
Gene ID (Entrez) 5241
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P06401-1
Quantity 100μL