missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: A21-W, Novus Biologicals™

$367.83 - $367.83

Specifications

Antigen Progesterone R/NR3C3
Clone A21-W
Immunogen Peptide derived from the region close to the N-terminus of human progesterone receptor.
Gene Accession No. P06401
Gene ID (Entrez) 5241
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17907810 Novus Biologicals Canada
NBP179078100UL
100μL Each for $367.83
Specifications Description & Specifications

Specifications

Progesterone R/NR3C3
Peptide derived from the region close to the N-terminus of human progesterone receptor.
5241
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
A21-W
P06401
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Primary
Rabbit
Unlabeled
Affinity Purified