Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: A21-W, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP179078200UL

 View all options for this product


For Research Use Only

Catalog No. NBP17907820


Description & Specifications


Antigen Progesterone R/NR3C3
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone A21-W
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Gene Accession No. P06401
Host Species Rabbit
Immunogen Peptide derived from the region close to the N-terminus of human progesterone receptor.
Isotype IgG
Purification Method Affinity Purified
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5241