Please login to your online account to display your discounted pricing

Rabbit anti-Progesterone R/NR3C3, Clone: A21-W, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179078100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907810

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Gene Accession No. P06401
Immunogen Peptide derived from the region close to the N-terminus of human progesterone receptor.
Quantity 100μL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Antigen Progesterone R/NR3C3
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgG
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 5241
Clone A21-W
Conjugate Unlabeled
Cross Reactivity Human