Please login to your online account to display your discounted pricing

Rabbit anti-PMEL17/SILV, Clone: P14-V, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179001100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17900110

  • / Each
 

Description & Specifications

Specifications

Antigen PMEL17/SILV
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone P14-V
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human melanosome/gp10.
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6490
Target Species Human