Please login to your online account to display your discounted pricing

Rabbit anti-PMEL17/SILV, Clone: P14-V, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179001200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17900120

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 200μL
Gene ID (Entrez) 6490
Regulatory Status RUO
Purification Method Affinity Purified
Conjugate Unlabeled
Antigen PMEL17/SILV
Immunogen Peptide derived from C-terminal sequence of human melanosome/gp10.
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Clone P14-V
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL