missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-PKC zeta, Clone: R14-K, Novus Biologicals™

$332.66 - $332.66

Specifications

Antigen PKC zeta
Clone R14-K
Concentration 1.5mg/mL
Immunogen Peptide derived from the human PKC-Zeta sequence.
Gene ID (Entrez) 5590
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP13012705 Novus Biologicals Canada
NBP130127005ML
0.05mL Each for $332.66
Specifications Description & Specifications

Specifications

PKC zeta
1.5mg/mL
5590
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Affinity Purified
ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
R14-K
Peptide derived from the human PKC-Zeta sequence.
20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Primary
Rabbit
Unlabeled
Affinity Purified