Please login to your online account to display your discounted pricing

Rabbit anti-PKC zeta, Clone: R14-K, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130127005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13012705

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen PKC zeta
Format Affinity Purified
Conjugate Unlabeled
Host Species Rabbit
Isotype IgG
Primary or Secondary Primary
Quantity 0.05mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone R14-K
Immunogen Peptide derived from the human PKC-Zeta sequence.
Test Specificity This antibody is specific for PKC-Zeta.
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Regulatory Status RUO
Purification Method Affinity Purified
Concentration 1.5mg/mL
Gene ID (Entrez) 5590