Please login to your online account to display your discounted pricing

Rabbit anti-PKC zeta, Clone: R14-K, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13012701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13012701

  • / Each
 

Description & Specifications

Specifications

Antigen PKC zeta
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Primary or Secondary Primary
Test Specificity This antibody is specific for PKC-Zeta.
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Host Species Rabbit
Immunogen Peptide derived from the human PKC-Zeta sequence.
Isotype IgG
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5590
Clone R14-K
Concentration 1.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat