Please login to your online account to display your discounted pricing

Rabbit anti-PKC gamma, Clone: A12-H, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen PKC gamma
Clone A12-H
Concentration 1.0mg/mL
Immunogen Peptide derived from the human PKC-gamma sequence
Gene Accession No. P05129
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13012405

 
novus biologicals canada
NBP130124005ML
0.05mL Each for $363.90
Description & Specifications

Specifications

Antigen PKC gamma
Clone A12-H
Concentration 1.0mg/mL
Immunogen Peptide derived from the human PKC-gamma sequence
Gene Accession No. P05129
Gene ID (Entrez) 5582
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting