Please login to your online account to display your discounted pricing

Rabbit anti-PKC gamma, Clone: A12-H, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130124005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13012405

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.05mL
Immunogen Peptide derived from the human PKC-gamma sequence
Gene Accession No. P05129
Antigen PKC gamma
Format Affinity Purified
Primary or Secondary Primary
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Host Species Rabbit
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 5582
Isotype IgG
Concentration 1.0mg/mL
Purification Method Affinity Purified
Clone A12-H
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is specific for PKC-gamma.