missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-PKC delta, Clone: K15-K, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen PKC delta
Clone K15-K
Concentration 0.5mg/mL
Immunogen Peptide derived from the human PKC-delta sequence
Gene Accession No. Q05655
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP13012505 Novus Biologicals Canada
NBP130125005ML
0.05mL Each for $363.90
Specifications Description & Specifications

Specifications

PKC delta
0.5mg/mL
Q05655
20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Primary
Rabbit
Unlabeled
Affinity Purified
K15-K
Peptide derived from the human PKC-delta sequence
5580
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Affinity Purified
ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot