Please login to your online account to display your discounted pricing

Rabbit anti-PKC delta, Clone: K15-K, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13012501ML

 View all options for this product


For Research Use Only

Catalog No. NBP13012501


Description & Specifications


Antigen PKC delta
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone K15-K
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Gene Accession No. Q05655
Host Species Rabbit
Immunogen Peptide derived from the human PKC-delta sequence
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5580
Test Specificity This antibody is specific for PKC-delta.