Please login to your online account to display your discounted pricing

Rabbit anti-Neprilysin/CD10, Clone: G27-P, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen Neprilysin/CD10
Clone G27-P
Immunogen Peptide derived from N-terminal region of human CD-10
Gene Accession No. P0847
Gene ID (Entrez) 4311
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17900310

 
novus biologicals canada
NBP179003100UL
100μL Each for $363.90
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone G27-P
Immunogen Peptide derived from N-terminal region of human CD-10
Gene Accession No. P0847
Gene ID (Entrez) 4311
Formulation 20mM Tris-HCl, pH 8. 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)