Please login to your online account to display your discounted pricing

Rabbit anti-Neprilysin/CD10, Clone: G27-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179003200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17900320

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Formulation 20mM Tris-HCl, pH 8. 20mg/mL BSA
Antigen Neprilysin/CD10
Immunogen Peptide derived from N-terminal region of human CD-10
Isotype IgG
Host Species Rabbit
Quantity 200μL
Gene ID (Entrez) 4311
Primary or Secondary Primary
Gene Accession No. P0847
Cross Reactivity Human
Clone G27-P
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled