Please login to your online account to display your discounted pricing

Rabbit anti-Neprilysin/CD10, Clone: E19-P, Novus Biologicals™

$332.66 - $483.00

Specifications

Antigen Neprilysin/CD10
Clone E19-P
Immunogen Peptide derived from human iNOS sequenc
Gene Accession No. P08473
Gene ID (Entrez) 4311
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17899610

 
novus biologicals canada
NBP178996100UL
100μL Each for $332.66

NBP17899620

 
novus biologicals canada
NBP178996200UL
200μL Each for $483.00
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone E19-P
Immunogen Peptide derived from human iNOS sequenc
Gene Accession No. P08473
Gene ID (Entrez) 4311
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)