Please login to your online account to display your discounted pricing

Rabbit anti-Neprilysin/CD10, Clone: E19-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178996200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17899620

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 200μL
Cross Reactivity Human
Antigen Neprilysin/CD10
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS.
Gene Accession No. P08473
Host Species Rabbit
Immunogen Peptide derived from human iNOS sequenc
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4311
Clone E19-P
Conjugate Unlabeled