Please login to your online account to display your discounted pricing

Rabbit anti-Nephrin, Clone: G17-H, Novus Biologicals™

$464.22 - $464.22

Specifications

Antigen Nephrin
Clone G17-H
Concentration 2.5mg/mL
Immunogen Peptide derived from the region of the first Ig-like domain of human nephrin (between residues Cys53-Cys101).
Gene Accession No. O60500
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13013705

 
novus biologicals canada
NBP130137005ML
0.05mL Each for $464.22
Description & Specifications

Specifications

Antigen Nephrin
Clone G17-H
Concentration 2.5mg/mL
Immunogen Peptide derived from the region of the first Ig-like domain of human nephrin (between residues Cys53-Cys101).
Gene Accession No. O60500
Gene ID (Entrez) 4868
Formulation 20mM Tris-HCl, pH 8.0. with 10mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Immunoprecipitation
Applications Western Blot