Please login to your online account to display your discounted pricing

Rabbit anti-Nephrin, Clone: G17-H, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13013701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13013701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunoprecipitation
Applications Western Blotting
Antigen Nephrin
Formulation 20mM Tris-HCl, pH 8.0. with 10mg/mL BSA. Immunogen affinity purified.
Gene Accession No. O60500
Isotype IgG
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Clone G17-H
Concentration 2.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Host Species Rabbit
Immunogen Peptide derived from the region of the first Ig-like domain of human nephrin (between residues Cys53-Cys101).
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4868
Test Specificity This antibody is specific for the C terminus of first Ig-like domain of human nephrin (between residues Cys53-Cys101).
Conjugate Unlabeled