Please login to your online account to display your discounted pricing

Rabbit anti-N-Cadherin, Clone: SP90, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide corresponding to C-terminus of human cadherin-N protein.
Regulatory Status RUO
Primary or Secondary Primary
Format Purified
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 1000
Test Specificity Predicted to cross react with Bovine, Mouse, Rat.
Reconstitution Protein A purified
Purification Method Purified
Clone SP90
Host Species Rabbit
Conjugate Unlabeled
Isotype IgG
Antigen N-Cadherin
Monoclonal or Polyclonal Monoclonal
Formulation PBS/1% BSA buffer pH 7.6
Cross Reactivity Human