Please login to your online account to display your discounted pricing

Rabbit anti-MUM1, Clone: SP114, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250505

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Primary or Secondary Primary
Regulatory Status RUO
Antigen MUM1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.5mL
Host Species Rabbit
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Formulation PBS/1% BSA buffer pH 7.6
Conjugate Unlabeled
Clone SP114
Reconstitution Protein A purified
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Mouse Rabbit, Rat.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 84939
Immunogen Synthetic peptide near C-terminus of human MUM1/IRF4.
Format Purified
Purification Method Purified