Please login to your online account to display your discounted pricing

Rabbit anti-MUM1, Clone: SP114, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250501

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 84939
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen MUM1
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide near C-terminus of human MUM1/IRF4.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Mouse Rabbit, Rat.
Reconstitution Protein A purified
Clone SP114
Conjugate Unlabeled
Cross Reactivity Human