missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-MS4A1/CD20, Clone: E17-P, Novus Biologicals™

$332.66 - $332.66

Specifications

Antigen MS4A1/CD20
Clone E17-P
Immunogen Peptide derived from C-terminal sequence of human CD20.
Gene Accession No. P11836
Gene ID (Entrez) 931
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP13014401 Novus Biologicals Canada
NBP13014401ML
0.1mL Each for $332.66
Specifications Description & Specifications

Specifications

MS4A1/CD20
Peptide derived from C-terminal sequence of human CD20.
931
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
E17-P
P11836
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary
Rabbit
Unlabeled
Affinity Purified