Please login to your online account to display your discounted pricing

Rabbit anti-MS4A1/CD20, Clone: E17-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Gene Accession No. P11836
Host Species Rabbit
Quantity 0.5mL
Conjugate Unlabeled
Antigen MS4A1/CD20
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Immunogen Peptide derived from C-terminal sequence of human CD20.
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 931
Test Specificity This antibody is specific for the C-terminus of CD20.
Clone E17-P
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.