Please login to your online account to display your discounted pricing

Rabbit anti-MS4A1/CD20, Clone: E17-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014401

  • / Each
 

Description & Specifications

Specifications

Antigen MS4A1/CD20
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone E17-P
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P11836
Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human CD20.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 931
Test Specificity This antibody is specific for the C-terminus of CD20.
Target Species Human