missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rabbit anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
Manufacturer: Mp Biomedicals 0855456
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Mouse
Specifications
IgG, Whole Molecule | |
Purified | |
IgG | |
Secondary |
Unconjugated | |
Rabbit | |
5mL | |
Polyclonal |