missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rabbit anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
$167.04 - $167.04
Specifications
Antigen | IgG, Whole Molecule |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN55436
|
MP Biomedicals Inc
0855436 |
2 mL |
Each for $167.04
|
|
|||||
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Mouse
Specifications
IgG, Whole Molecule | |
Unconjugated | |
Rabbit | |
Secondary |
Polyclonal | |
Purified | |
IgG |