Please login to your online account to display your discounted pricing

Rabbit anti-Mesothelin, Clone: SP74, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen Mesothelin
Clone SP74
Immunogen Synthetic peptide corresponding to C-terminus of human mesothelin protein.
Gene ID (Entrez) 10232
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21248301

 
novus biologicals canada
NBP21248301ML
0.1mL Each for $309.05

NBP21248305

 
novus biologicals canada
NBP21248305ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen Mesothelin
Clone SP74
Immunogen Synthetic peptide corresponding to C-terminus of human mesothelin protein.
Gene ID (Entrez) 10232
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)