Please login to your online account to display your discounted pricing

Rabbit anti-Mesothelin, Clone: SP74, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248305

  • / Each
 

Description & Specifications

Specifications

Antigen Mesothelin
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human mesothelin protein.
Isotype IgG
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10232
Conjugate Unlabeled
Format Purified
Purification Method Purified
Regulatory Status RUO
Test Specificity Predicted to cross react with Chicken, Porcine.
Reconstitution Protein A purified
Clone SP74